missing translation for 'onlineSavingsMsg'
Learn More

4-(1,2,4-Triazol-1-ylmethyl)aniline 98.0+%, TCI America™

Catalog No. p-7117760
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21481G 1 g
A21485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21481G Supplier TCI America Supplier No. A21481G

Chemical Identifiers

CAS 119192-10-8
Molecular Formula C9H10N4
Molecular Weight (g/mol) 174.207
MDL Number MFCD00973306
InChI Key ZGLQVRIVLWGDNA-UHFFFAOYSA-N
Synonym 4-1h-1,2,4-triazol-1-ylmethyl aniline, 4-1h-1,2,4-triazol-1-yl methyl aniline, 1-4-aminobenzyl-1,2,4-triazole, 4-1,2,4-triazol-1-ylmethyl aniline, 4-1h-1,2,4-triazol-1-yl-methyl benzenamine, 4-1,2,4 triazol-1-ylmethylphenylamine, benzenamine, 4-1h-1,2,4-triazol-1-ylmethyl, 4-1,2,4 triazol-1-ylmethyl-phenylamine, 4-1h-1,2,4-triazol-1-yl-methyl aniline, 4-1h-1,2,4-triazol-1-ylmethyl benzenamine
PubChem CID 821219
IUPAC Name 4-(1,2,4-triazol-1-ylmethyl)aniline
SMILES C1=CC(=CC=C1CN2C=NC=N2)N

Specifications

Melting Point 128°C
Color White-Yellow
Quantity 1 g
Formula Weight 174.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(1,2,4-Triazol-1-ylmethyl)aniline