missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1,2,4-Triazol-1-yl)benzaldehyde 98.0+%, TCI America™
$296.38 - $1051.33
Chemical Identifiers
| CAS | 27996-86-7 |
|---|---|
| Molecular Formula | C9H7N3O |
| Molecular Weight (g/mol) | 173.175 |
| MDL Number | MFCD02681969 |
| InChI Key | TVEJNWMWDIXPAX-UHFFFAOYSA-N |
| PubChem CID | 2776488 |
| IUPAC Name | 4-(1,2,4-triazol-1-yl)benzaldehyde |
| SMILES | C1=CC(=CC=C1C=O)N2C=NC=N2 |
Chemical Identifiers
| 27996-86-7 | |
| 173.175 | |
| TVEJNWMWDIXPAX-UHFFFAOYSA-N | |
| 4-(1,2,4-triazol-1-yl)benzaldehyde |
| C9H7N3O | |
| MFCD02681969 | |
| 2776488 | |
| C1=CC(=CC=C1C=O)N2C=NC=N2 |
Specifications
| 27996-86-7 | |
| Yellow | |
| MFCD02681969 | |
| 4-1h-1,2,4-triazol-1-yl benzaldehyde, 4-1,2,4 triazol-1-yl-benzaldehyde, 4-1,2,4-triazol-1-yl benzaldehyde, 4-1h-1,2,4-triazol-1-yl benzenecarbaldehyde, benzaldehyde, 4-1h-1,2,4-triazol-1-yl, 1-4-formylphenyl-1,2,4-triazole, benzaldehyde,4-1h-1,2,4-triazol-1-yl, 4-1,2,4-triazolyl benzaldehyde, 4-1,2,4-triazol-1-yl-benzaldehyde | |
| C1=CC(=CC=C1C=O)N2C=NC=N2 | |
| 173.175 | |
| 173.18 | |
| Crystalline Powder |
| 149°C | |
| C9H7N3O | |
| 1 g | |
| TVEJNWMWDIXPAX-UHFFFAOYSA-N | |
| 4-(1,2,4-triazol-1-yl)benzaldehyde | |
| 2776488 | |
| ≥98.0% (GC,T) | |
| 4-(1,2,4-Triazol-1-yl)benzaldehyde |
Safety and Handling
TSCA : No