CAS RN 27996-86-7
IUPAC Name:
4-(1H-1,2,4-triazol-1-yl)benzaldehyde
Synonyms:
4-(1,2,4-triazol-1-yl)benzaldehyde
Molecular Weight (g/mol):
173.18
Molecular Formula:
C9H7N3O
InChi Key:
TVEJNWMWDIXPAX-UHFFFAOYSA-N
SMILES:
O=CC1=CC=C(C=C1)N1C=NC=N1
1
–
1
of
1
results
1
4-(1,2,4-Triazol-1-yl)benzaldehyde 98.0+%, TCI America™
CAS: 27996-86-7 Molecular Formula: C9H7N3O Molecular Weight (g/mol): 173.175 MDL Number: MFCD02681969 InChI Key: TVEJNWMWDIXPAX-UHFFFAOYSA-N PubChem CID: 2776488 IUPAC Name: 4-(1,2,4-triazol-1-yl)benzaldehyde SMILES: C1=CC(=CC=C1C=O)N2C=NC=N2