missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1,2,4-Triazol-1-yl)benzaldehyde 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 149°C |
| Color | Yellow |
| Quantity | 5 g |
| Synonym | 4-1h-1,2,4-triazol-1-yl benzaldehyde, 4-1,2,4 triazol-1-yl-benzaldehyde, 4-1,2,4-triazol-1-yl benzaldehyde, 4-1h-1,2,4-triazol-1-yl benzenecarbaldehyde, benzaldehyde, 4-1h-1,2,4-triazol-1-yl, 1-4-formylphenyl-1,2,4-triazole, benzaldehyde,4-1h-1,2,4-triazol-1-yl, 4-1,2,4-triazolyl benzaldehyde, 4-1,2,4-triazol-1-yl-benzaldehyde |
| Formula Weight | 173.18 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-(1,2,4-Triazol-1-yl)benzaldehyde |
Safety and Handling
| TSCA | No |