missing translation for 'onlineSavingsMsg'
Learn More

4-(1,2,4-Triazol-1-yl)benzaldehyde 98.0+%, TCI America™

Catalog No. F09031G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09031G 1 g
F09035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09031G Supplier TCI America Supplier No. F09031G
Only null left

Chemical Identifiers

CAS 27996-86-7
Molecular Formula C9H7N3O
Molecular Weight (g/mol) 173.175
MDL Number MFCD02681969
InChI Key TVEJNWMWDIXPAX-UHFFFAOYSA-N
PubChem CID 2776488
IUPAC Name 4-(1,2,4-triazol-1-yl)benzaldehyde
SMILES C1=CC(=CC=C1C=O)N2C=NC=N2

Specifications

Melting Point 149°C
Color Yellow
Quantity 1 g
Synonym 4-1h-1,2,4-triazol-1-yl benzaldehyde, 4-1,2,4 triazol-1-yl-benzaldehyde, 4-1,2,4-triazol-1-yl benzaldehyde, 4-1h-1,2,4-triazol-1-yl benzenecarbaldehyde, benzaldehyde, 4-1h-1,2,4-triazol-1-yl, 1-4-formylphenyl-1,2,4-triazole, benzaldehyde,4-1h-1,2,4-triazol-1-yl, 4-1,2,4-triazolyl benzaldehyde, 4-1,2,4-triazol-1-yl-benzaldehyde
Formula Weight 173.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(1,2,4-Triazol-1-yl)benzaldehyde