missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1,2,4-Triazol-1-yl)aniline 98.0+%, TCI America™
Supplier: TCI America T30131G
Specifications
| 4-(1,2,4-Triazol-1-yl)aniline | |
| 143°C | |
| MFCD00171470 | |
| 4-1,2,4 triazol-1-yl-phenylamine, 4-1h-1,2,4-triazol-1-yl aniline, 4-1,2,4-triazol-1-yl aniline, 1-4-aminophenyl-1,2,4-triazole, triazolylaniline, 1-4'-aminophenyl-1,2,4-triazole, benzenamine, 4-1h-1,2,4-triazol-1-yl, 4-1h-1,2,4-triazol-1-yl benzenamine, 4-1,2,4-triazolyl phenylamine, pubchem3927 | |
| C1=CC(=CC=C1N)N2C=NC=N2 | |
| 160.18 | |
| 160.18 | |
| Crystalline Powder |
| 6523-49-5 | |
| C8H8N4 | |
| 1 g | |
| BLNPEJXSNSBBNM-UHFFFAOYSA-N | |
| 4-(1,2,4-triazol-1-yl)aniline | |
| 735826 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 6523-49-5 | |
| 160.18 | |
| BLNPEJXSNSBBNM-UHFFFAOYSA-N | |
| 735826 | |
| C1=CC(=CC=C1N)N2C=NC=N2 |
| C8H8N4 | |
| MFCD00171470 | |
| 4-1,2,4 triazol-1-yl-phenylamine, 4-1h-1,2,4-triazol-1-yl aniline, 4-1,2,4-triazol-1-yl aniline, 1-4-aminophenyl-1,2,4-triazole, triazolylaniline, 1-4'-aminophenyl-1,2,4-triazole, benzenamine, 4-1h-1,2,4-triazol-1-yl, 4-1h-1,2,4-triazol-1-yl benzenamine, 4-1,2,4-triazolyl phenylamine, pubchem3927 | |
| 4-(1,2,4-triazol-1-yl)aniline |
Safety and Handling
TSCA : No