missing translation for 'onlineSavingsMsg'
Learn More

4-(1,2,4-Triazol-1-yl)aniline 98.0+%, TCI America™

Catalog No. p-7137529
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3013200MG 200 mg
T30131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3013200MG Supplier TCI America Supplier No. T3013200MG
Only null left

Chemical Identifiers

CAS 6523-49-5
Molecular Formula C8H8N4
Molecular Weight (g/mol) 160.18
MDL Number MFCD00171470
InChI Key BLNPEJXSNSBBNM-UHFFFAOYSA-N
Synonym 4-1,2,4 triazol-1-yl-phenylamine, 4-1h-1,2,4-triazol-1-yl aniline, 4-1,2,4-triazol-1-yl aniline, 1-4-aminophenyl-1,2,4-triazole, triazolylaniline, 1-4'-aminophenyl-1,2,4-triazole, benzenamine, 4-1h-1,2,4-triazol-1-yl, 4-1h-1,2,4-triazol-1-yl benzenamine, 4-1,2,4-triazolyl phenylamine, pubchem3927
PubChem CID 735826
IUPAC Name 4-(1,2,4-triazol-1-yl)aniline
SMILES C1=CC(=CC=C1N)N2C=NC=N2

Specifications

Melting Point 143°C
Quantity 200 mg
Formula Weight 160.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(1,2,4-Triazol-1-yl)aniline