missing translation for 'onlineSavingsMsg'
Learn More

4-(1,1,2,2-Tetrafluoroethoxy)benzoic Acid 98.0+%, TCI America™

$121.16 - $367.22

Chemical Identifiers

CAS 10009-25-3
Molecular Formula C9H6F4O3
Molecular Weight (g/mol) 238.14
MDL Number MFCD00155926
InChI Key SVGWTILJZWYEMD-UHFFFAOYSA-N
PubChem CID 2776639
IUPAC Name 4-(1,1,2,2-tetrafluoroethoxy)benzoic acid
SMILES OC(=O)C1=CC=C(OC(F)(F)C(F)F)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T30275G
View Documents
TCI America
T30275G
5 g
Each for $121.16
Only null left
 
T302725G
View Documents
TCI America
T302725G
25 g
Each for $367.22
Only null left
 
Description

Description

Specifications

Chemical Identifiers

10009-25-3
238.14
SVGWTILJZWYEMD-UHFFFAOYSA-N
4-(1,1,2,2-tetrafluoroethoxy)benzoic acid
C9H6F4O3
MFCD00155926
2776639
OC(=O)C1=CC=C(OC(F)(F)C(F)F)C=C1

Specifications

10009-25-3
C9H6F4O3
5 g
OC(=O)C1=CC=C(OC(F)(F)C(F)F)C=C1
238.14
238.14
Crystalline Powder
178°C
MFCD00155926
SVGWTILJZWYEMD-UHFFFAOYSA-N
4-(1,1,2,2-tetrafluoroethoxy)benzoic acid
2776639
≥98.0% (GC,T)
4-(1,1,2,2-Tetrafluoroethoxy)benzoic Acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents