missing translation for 'onlineSavingsMsg'
Learn More

4-(1,1,2,2-Tetrafluoroethoxy)benzaldehyde 97.0+%, TCI America™

Catalog No. T24815G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24815G 5 g
T24811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24815G Supplier TCI America Supplier No. T24815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35295-36-4
Molecular Formula C9H6F4O2
Molecular Weight (g/mol) 222.14
MDL Number MFCD00039577
InChI Key ZTBIQWAGWYPSHC-UHFFFAOYSA-N
PubChem CID 118804
IUPAC Name 4-(1,1,2,2-tetrafluoroethoxy)benzaldehyde
SMILES FC(F)C(F)(F)OC1=CC=C(C=O)C=C1

Specifications

Melting Point 9°C
Color Yellow
Boiling Point 103°C
Quantity 5 g
Formula Weight 222.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(1,1,2,2-Tetrafluoroethoxy)benzaldehyde