accessibility menu, dialog, popup

UserName

CAS RN 35295-36-4

IUPAC Name: 4-(1,1,2,2-tetrafluoroethoxy)benzaldehyde
Synonyms: 4-(1,1,2,2-tetrafluoroethoxy)benzaldehyde
Molecular Weight (g/mol): 222.14
Molecular Formula: C9H6F4O2
InChi Key: ZTBIQWAGWYPSHC-UHFFFAOYSA-N
SMILES: FC(F)C(F)(F)OC1=CC=C(C=O)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results