missing translation for 'onlineSavingsMsg'
Learn More

(3S)-(+)-1-Benzyl-3-(ethylamino)pyrrolidine 95.0+%, TCI America™

Catalog No. B15815G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B15815G Supplier TCI America Supplier No. B15815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 169750-99-6
Molecular Formula C13H20N2
Molecular Weight (g/mol) 204.32
MDL Number MFCD00191307
InChI Key ZSIUSRJKSLXIJH-UHFFFAOYNA-N
PubChem CID 12362914
IUPAC Name 1-benzyl-N-ethylpyrrolidin-3-amine
SMILES CCNC1CCN(CC2=CC=CC=C2)C1

Specifications

Color Yellow
Quantity 5 g
UN Number 2735
Formula Weight 204.32
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (3S)-(+)-1-Benzyl-3-(ethylamino)pyrrolidine