missing translation for 'onlineSavingsMsg'
Learn More

(3S)-(+)-1-Benzyl-3-aminopyrrolidine 98.0+%, TCI America™

Catalog No. A117410G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A117410G 10 g
A117425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A117410G Supplier TCI America Supplier No. A117410G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 114715-38-7
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.263
MDL Number MFCD00082766
InChI Key HBVNLKQGRZPGRP-NSHDSACASA-N
Synonym s-+-1-benzyl-3-aminopyrrolidine, 3s-1-benzylpyrrolidin-3-amine, 3s-+-1-benzyl-3-aminopyrrolidine, s-n-benzyl-3-aminopyrrolidine, s-3-amino-1-n-benzyl-pyrrolidine, s-1-benzylpyrrolidin-3-amine, s-1-benzyl-3-aminopyrrolidine, s-3-amino-1-benzylpyrrolidine, s-bap, s-3-amino-1-n-benzylpyrrolidine
PubChem CID 1519353
IUPAC Name (3S)-1-benzylpyrrolidin-3-amine
SMILES C1CN(CC1N)CC2=CC=CC=C2

Specifications

Color Yellow
Quantity 10 g
UN Number 2735
Formula Weight 176.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (3S)-(+)-1-Benzyl-3-aminopyrrolidine