missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3R,4S)-3-Hydroxy-4-phenyl-2-azetidinone 98.0+%, TCI America™
Supplier: TCI America H16561G
Specifications
| (3R,4S)-3-Hydroxy-4-phenyl-2-azetidinone | |
| 188°C | |
| C9H9NO2 | |
| 1 g | |
| C1=CC=C(C=C1)C2C(C(=O)N2)O | |
| 163.176 | |
| 163.18 | |
| Crystalline Powder |
| 132127-34-5 | |
| White-Yellow | |
| MFCD07368374 | |
| FBZSDKXFQUKDLD-JGVFFNPUSA-N | |
| (3R,4S)-3-hydroxy-4-phenylazetidin-2-one | |
| 10219589 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 132127-34-5 | |
| 163.176 | |
| FBZSDKXFQUKDLD-JGVFFNPUSA-N | |
| (3R,4S)-3-hydroxy-4-phenylazetidin-2-one |
| C9H9NO2 | |
| MFCD07368374 | |
| 10219589 | |
| C1=CC=C(C=C1)C2C(C(=O)N2)O |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator