accessibility menu, dialog, popup

UserName

CAS RN 132127-34-5

OHNOH

CAS RN 132127-34-5

IUPAC Name: 3-hydroxy-4-phenylazetidin-2-one
Synonyms: 3-hydroxy-4-phenylazetidin-2-one
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: FBZSDKXFQUKDLD-UHFFFAOYNA-N
SMILES: OC1C(NC1=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results