missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine 98.0+%, TCI America™
Supplier: TCI America A117125G
| Quantity | 25 g |
|---|
Chemical Identifiers
| 122536-77-0 | |
| 186.255 | |
| DQQJBEAXSOOCPG-SSDOTTSWSA-N | |
| 1514397 | |
| CC(C)(C)OC(=O)NC1CCNC1 |
| C9H18N2O2 | |
| MFCD00143191 | |
| r-3-boc-amino pyrrolidine, r-tert-butyl pyrrolidin-3-ylcarbamate, r-+-3-boc-amino pyrrolidine, tert-butyl n-3r-pyrrolidin-3-yl carbamate, 3r-+-3-tert-butoxycarbonylamino pyrrolidine, r-3-n-boc-aminopyrrolidine, 3r-+-3-boc-amino pyrrolidine, r-3-boc-aminopyrrolidine, tert-butyl r-pyrrolidin-3-ylcarbamate, tert-butyl 3r-pyrrolidin-3-ylcarbamate | |
| tert-butyl N-[(3R)-pyrrolidin-3-yl]carbamate |
Specifications
| (3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine | |
| C9H18N2O2 | |
| r-3-boc-amino pyrrolidine, r-tert-butyl pyrrolidin-3-ylcarbamate, r-+-3-boc-amino pyrrolidine, tert-butyl n-3r-pyrrolidin-3-yl carbamate, 3r-+-3-tert-butoxycarbonylamino pyrrolidine, r-3-n-boc-aminopyrrolidine, 3r-+-3-boc-amino pyrrolidine, r-3-boc-aminopyrrolidine, tert-butyl r-pyrrolidin-3-ylcarbamate, tert-butyl 3r-pyrrolidin-3-ylcarbamate | |
| CC(C)(C)OC(=O)NC1CCNC1 | |
| 25 g | |
| 1514397 | |
| ≥98.0% (GC,T) |
| 122536-77-0 | |
| MFCD00143191 | |
| DQQJBEAXSOOCPG-SSDOTTSWSA-N | |
| tert-butyl N-[(3R)-pyrrolidin-3-yl]carbamate | |
| 186.255 | |
| 186.26 | |
| Crystalline Powder |
Safety and Handling
TSCA : No