missing translation for 'onlineSavingsMsg'
Learn More

(3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine 98.0+%, TCI America™

Catalog No. p-7117283
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11715G 5 g
A117125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A11715G Supplier TCI America Supplier No. A11715G

Chemical Identifiers

CAS 122536-77-0
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.255
MDL Number MFCD00143191
InChI Key DQQJBEAXSOOCPG-SSDOTTSWSA-N
Synonym r-3-boc-amino pyrrolidine, r-tert-butyl pyrrolidin-3-ylcarbamate, r-+-3-boc-amino pyrrolidine, tert-butyl n-3r-pyrrolidin-3-yl carbamate, 3r-+-3-tert-butoxycarbonylamino pyrrolidine, r-3-n-boc-aminopyrrolidine, 3r-+-3-boc-amino pyrrolidine, r-3-boc-aminopyrrolidine, tert-butyl r-pyrrolidin-3-ylcarbamate, tert-butyl 3r-pyrrolidin-3-ylcarbamate
PubChem CID 1514397
IUPAC Name tert-butyl N-[(3R)-pyrrolidin-3-yl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCNC1

Specifications

Quantity 5 g
Formula Weight 186.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine