missing translation for 'onlineSavingsMsg'
Learn More

[(3R)-3-Hydroxytetradecanoyl]-L-carnitine, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013150
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013150 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013150 Supplier Merck Emd Millipore Supplier No. 5200210MG

Specifications

CAS 1469901-02-7
Linear Formula C21H41 NO5
Synonym (2 R)-3-Carboxy-2-[[(3 R)-3-hydroxy-1-oxotetradecyl]oxy]-N,N,N-trimethyl-1-propanaminium inner salt
Type Analytical Standard
Quantity 10 mg
Formula Weight 387.55
Percent Purity ≥90% (HPLC)
Grade Analytical Standard