missing translation for 'onlineSavingsMsg'
Learn More

(3R)-(+)-1-Benzyl-3-(tert-butoxycarbonylamino)pyrrolidine 98.0+%, TCI America™

Catalog No. B19325G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B19325G Supplier TCI America Supplier No. B19325G
Only null left

Chemical Identifiers

CAS 131878-23-4
Molecular Formula C16H24N2O2
Molecular Weight (g/mol) 276.38
MDL Number MFCD01321148
InChI Key PHOIDJGLYWEUEK-CQSZACIVSA-N
PubChem CID 10286119
IUPAC Name tert-butyl N-[(3R)-1-benzylpyrrolidin-3-yl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCN(C1)CC2=CC=CC=C2

Specifications

Melting Point 80°C
Color White
Quantity 5 g
Formula Weight 276.38
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (3R)-(+)-1-Benzyl-3-(tert-butoxycarbonylamino)pyrrolidine