missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Standard Chemical Sorbents
Description
Meltblown polypropylene microfibers absorb liquids such as hydrofluoric acid and other aqueous-based chemicals, and soak up an average of 10 times their weight of most EPA-designated hazardous liquids.
- Sorbents are dirt- and dust-free; contain no abrasive granules that can damage expensive equipment
- Polypropylene base is chemically inert and incinerable; allows separation and recycling in many applications
- Easy-to-spot yellow cartons protect sorbents from temperature and humidity for secure, long-term storage
- Convenient dispenser boxes—suitable for tabletops or wall mounting—keep pads and pillows within easy reach Pads or rolls are ideal for use on work surfaces and can be trimmed to size
- Mini booms surround and contain spills; pillows soak them up
- 3M Catalog# P110, P200, P208, P300, P500
These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
Specifications
Specifications
| Certifications/Compliance | OSHA 29 CFR 1910.132 |
| Color | Yellow |
| Capacity (English) | 8 gal. |
| Capacity (Metric) | 30.3 L |
| Material | Polypropylene |
| Type | Chemical Sorbent Pillow |
| For Use With (Application) | For control of spills of certain hazardous liquids |
| Model | P300 |
| Dimensions (L x W) | 38 x 17.78 cm (15 x 7 in.) |
| Length (English) | 15 in. |
| Show More |