missing translation for 'onlineSavingsMsg'
Learn More

3M™ Standard Chemical Sorbents

Catalog No. 172102
Change view
Click to view available options
Capacity (English):
8 gal.
12 gal.
17 gal.
Type:
Chemical Sorbent Mini Boom
Chemical Sorbent Pad
Chemical Sorbent Pillow
Model:
P110
P200
P208
P300
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Type Model
172102 12 gal. Chemical Sorbent Mini Boom P200
17210 17 gal. Chemical Sorbent Pad P110
172103 8 gal. Chemical Sorbent Pillow P300
19039448 12 gal. Chemical Sorbent Mini Boom P208
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 172102 Supplier 3M™ Supplier No. 7000001909

Absorb virtually any liquid with these chemical sorbents

Meltblown polypropylene microfibers absorb liquids such as hydrofluoric acid and other aqueous-based chemicals, and soak up an average of 10 times their weight of most EPA-designated hazardous liquids.

  • Sorbents are dirt- and dust-free; contain no abrasive granules that can damage expensive equipment
  • Polypropylene base is chemically inert and incinerable; allows separation and recycling in many applications
  • Easy-to-spot yellow cartons protect sorbents from temperature and humidity for secure, long-term storage
  • Convenient dispenser boxes—suitable for tabletops or wall mounting—keep pads and pillows within easy reach Pads or rolls are ideal for use on work surfaces and can be trimmed to size
  • Mini booms surround and contain spills; pillows soak them up
  • 3M Catalog# P110, P200, P208, P300, P500

These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.

Specifications

Certifications/Compliance OSHA 29 CFR 1910.132
Color Yellow
Capacity (English) 12 gal.
Capacity (Metric) 45.4 L
Material Polypropylene
Type Chemical Sorbent Mini Boom
For Use With (Application) For control of spills of certain hazardous liquids
Model P200
Length (English) 4 ft.
Length (Metric) 1.21 m
Diameter (English) 3 in.
Diameter (Metric) 7.62 cm
Show More Show Less