Learn More
3M™ L-Series Headgear Accessories
For use with various 3M™ L-Series headgear.
$59.05 - $167.63
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
19-038-947
|
3M™
7000126263 |
1 Case |
Each for $59.05
|
|
|||||
|
19-038-932
|
3M™
7000126255 |
2/Cs. |
Pack of 2 Each for $167.63
|
|
|||||
Description
3M™ offers a variety of accessories for L-Series Headgear- L-129-2: Used with the 3M™ Headgear L-Series (except the L-900 Series Headgear). It is attached to the headgear by two matching 3M™ Scotchmate™ Hook-and-Loop Fasteners and drapes over the worker for safety purposes.
- L-140-10: Used on the 3M™ Wide View face shield L-130. The face shield attaches to the headgear using the positioning cam mechanism that is secured by the knob to each side of the headgear.
- L-140-10: Used on the 3M™ Welding face shield and Combination face shield. The kit includes the 3M™ Welding Knob L-145 and the pivot post on which the face shield pivots when raised, lowered and repositioned.
- 3M Catalog# L-129-2 , 3M Catalog# L-140-10 , 3M Catalog# L-146