Learn More
3M™ L-Series Headgear Accessories
For use with various 3M™ L-Series headgear.
Supplier:  3M™ 7000126263
 View more versions of this product
Catalog No. 19-038-947
Only null left
Explore available promotions
Description
Description
3M™ offers a variety of accessories for L-Series Headgear
  • L-129-2: Used with the 3M™ Headgear L-Series (except the L-900 Series Headgear). It is attached to the headgear by two matching 3M™ Scotchmate™ Hook-and-Loop Fasteners and drapes over the worker for safety purposes.
  • L-140-10: Used on the 3M™ Wide View face shield L-130. The face shield attaches to the headgear using the positioning cam mechanism that is secured by the knob to each side of the headgear.
  • L-140-10: Used on the 3M™ Welding face shield and Combination face shield. The kit includes the 3M™ Welding Knob L-145 and the pivot post on which the face shield pivots when raised, lowered and repositioned.
  • 3M Catalog# L-129-2 , 3M Catalog# L-140-10 , 3M Catalog# L-146
Specifications
Specifications
3M Headgear L-Series
3M™ Spring loaded welding knob L-145 and pivot post
Welding Knob and Pivot Kit
For grinding, machine operation, metal production etc
L-146
1 Case
Product Suggestions
Product Suggestions