missing translation for 'onlineSavingsMsg'
Learn More

3M KCl AgCl Electrolyte Solution, METTLER TOLEDO™

Catalog No. 01912584
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01912584 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01912584 Supplier METTLER TOLEDO Supplier No. 51350074/DUP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Maintenance and pH Electrolyte Solution

  • 3M KCl Saturated AgCl
  • pH Electrolyte and Maintenance Solution
  • 250 mL

Specifications

Chemical Name or Material Electrolyte, KCL Saturated AgCl
Color Colorless
CAS 7447-40-7
Boiling Point 1420°C
Melting Point 770°C
Molecular Formula KCl, AgCl
Concentration 3 M
Synonym None
Molecular Weight (g/mol) 74.55
Packaging Plastic Bottle
pH ∼6
Quantity 250 mL
Show More Show Less