missing translation for 'onlineSavingsMsg'
Learn More

3M™ High-Capacity Chemical Sorbents

Catalog No. p-37623
Change view
Click to view available options
Capacity (English):
10.5 gal.
23.5 gal.
31 gal.
Length (English):
9.05 in.
50 ft.
150 ft.
Width (English):
5 in.
14.17 in.
15.25 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Length (English) Width (English)
18999564 31 gal. 150 ft. 15.25 in.
19083193 23.5 gal. 9.05 in. 14.17 in.
19087815 10.5 gal. 50 ft. 5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18999564 Supplier 3M™ Supplier No. 7000001948
Only null left

Made from inert polypropylene and polyester fibers

  • Suitable for a wide range of liquids
  • Lightweight, easy to deploy and dust free
  • Identifiable coloring for easy identification and visibility
  • Can be wrung out and reused
  • High absorption capacity minimizes amount of waste for disposal
  • L x W: 50 ft. x 5 in. (15.2m x 12.7cm)
  • Sorbing Capacity: 31.5 gal. (119.2L)
  • 3M Catalog# CFL550DD , 3M Catalog# CRL15150 , 3M Catalog# PD914

These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.

Specifications

Certifications/Compliance OSHA 29 CFR 1910.132
Color Yellow
Capacity (English) 31 gal.
Capacity (Metric) 117.3 L
Material Polyester
Type Chemical Sorbent Roll
For Use With (Application) For control of spills of certain hazardous liquids
Model CRL15150
Dimensions (L x W) 45.7 m x 38.73 cm (150 ft. x 15.25 in.)
Length (English) 150 ft.
Length (Metric) 45.7 m
Width (English) 15.25 in.
Width (Metric) 38.73 cm
Show More Show Less