Learn More
3M™ BX™ Readers Protective Eyewear
Dual diopter available
Supplier: 3M™ 7000127493
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Description
- High-impact polycarbonate lens absorbs 99.9% UV
- Adjustable soft-tip temples feel comfortable even after extended wear
- Soft nose bridge
- Three lens angles for custom fit
- 3M Catalog# 11374-00000-20, 11375-00000-20, 11376-00000-20, 11377-00000-20, 11378-00000-20, 11379-00000-20, 11457-00000-20, 11458-00000-20, 11475-00000-20, 11473-00000-20, 11474-00000-20
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
Specifications
| Scratch-resistant | |
| Gray | |
| Silver | |
| 11378-00000-20 |
| ANSI Z87.1-2015, CSA Z94.3 | |
| Reader Protective Eyewear | |
| +2.00 | |
| UV400 |