missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Protective Coverall 4540+, Hooded
Coverall is made from a laminated premium bicomponent non-woven and features a breathable SMMMS triangular back panel for improved comfort. Soft and lightweight material provides an ideal barrier to hazardous particles and limited liquid splashes.
$282.53 - $282.53
Specifications
| Material | Polyethylene, Polyester/Polyethylene Laminate |
|---|---|
| Color | White |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
17-623-357
|
3M™
7100041597 |
3X-Large |
Case of 25 Each for $282.53
|
|
|||||
Description
- Ideal barrier against dry particles and certain liquid chemical splash (CE Type 5/6)
- Soft and light, ultra low-linting laminated microporous material that drapes and hangs well over the body facilitating ease of movement
- 3 panel hood, 2-way zip and storm flap with sealable tape with elasticated back and ankles to reduce the risk of skin exposure
- Knitted cuffs and elasticated waist and ankles for increased comfort and freedom of movement
- Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
- EN 1073-2 nuclear particulate hold out
Specifications
| Polyethylene, Polyester/Polyethylene Laminate | |
| Single-use | |
| Elastic | |
| 3-panel | |
| Anti-static |
| White | |
| Elastic | |
| Hooded | |
| Serged, Bound | |
| White Polypropylene/Polyethylene laminated suit, SMMMS Polypropylene blue back panel, Nylon on Polyester braid zipper, Neoprene Rubber elastic, Polyester cuffs and thread |