missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3aR)-(+)-Sclareolide 97.0+%, TCI America™
Supplier: TCI America S084725G
Spécifications
| (3aR)-(+)-Sclareolide | |
| 124°C | |
| C16H26O2 | |
| 25 g | |
| CC12CCC3C(C)(C)CCCC3(C)C1CC(=O)O2 | |
| 250.38 | |
| 250.38 | |
| Crystalline Powder |
| 564-20-5 | |
| White | |
| MFCD00134168 | |
| IMKJGXCIJJXALX-UHFFFAOYNA-N | |
| 3a,6,6,9a-tetramethyl-dodecahydronaphtho[2,1-b]furan-2-one | |
| 929262 | |
| ≥97.0% (GC) |
Identifiants chimiques
| 564-20-5 | |
| 250.38 | |
| IMKJGXCIJJXALX-UHFFFAOYNA-N | |
| 3a,6,6,9a-tetramethyl-dodecahydronaphtho[2,1-b]furan-2-one |
| C16H26O2 | |
| MFCD00134168 | |
| 929262 | |
| CC12CCC3C(C)(C)CCCC3(C)C1CC(=O)O2 |
Sécurité et manipulation
missing translation for 'tsca' : Yes