accessibility menu, dialog, popup

UserName

CH3H3CH3CH3COO

CAS RN 564-20-5

IUPAC Name: 3a,6,6,9a-tetramethyl-dodecahydronaphtho[2,1-b]furan-2-one
Synonyms: sclareolide
Molecular Weight (g/mol): 250.38
Molecular Formula: C16H26O2
InChi Key: IMKJGXCIJJXALX-UHFFFAOYNA-N
SMILES: CC12CCC3C(C)(C)CCCC3(C)C1CC(=O)O2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results