missing translation for 'onlineSavingsMsg'
Learn More

(3aR)-(+)-Sclareolide 97.0+%, TCI America™

Catalog No. S08475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08475G 5 g
S084725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S08475G Supplier TCI America Supplier No. S08475G

Chemical Identifiers

CAS 564-20-5
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00134168
InChI Key IMKJGXCIJJXALX-UHFFFAOYNA-N
PubChem CID 929262
IUPAC Name 3a,6,6,9a-tetramethyl-dodecahydronaphtho[2,1-b]furan-2-one
SMILES CC12CCC3C(C)(C)CCCC3(C)C1CC(=O)O2

Specifications

Melting Point 124°C
Color White
Quantity 5 g
Formula Weight 250.38
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (3aR)-(+)-Sclareolide