missing translation for 'onlineSavingsMsg'
Learn More

(3aR,4S,5R,6aS)-(-)-Hexahydro-5-hydroxy-4-hydroxymethyl-2H-cyclopenta[b]furan-2-one, 98%

Catalog No. p-7037396
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH56119MD 250 mg
AAH5611903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH56119MD Supplier Thermo Scientific Chemicals Supplier No. H56119MD
Only null left

CAS: 32233-40-2 | C8H12O4 | 172.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32233-40-2
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00075234
InChI Key VYTZWRCSPHQSFX-GBNDHIKLSA-N
Synonym --corey lactone diol, 3ar,4s,5r,6as-5-hydroxy-4-hydroxymethyl hexahydro-2h-cyclopenta b furan-2-one, --corey lactone, corey lactone diol, --corey diol corey lactone, 3ar,4s,5r,6as---hexahydro-5-hydroxy-4-hydroxymethyl-2h-cyclopenta b furan-2-one, 1s,6s,5r,7r-7-hydroxy-6-hydroxymethyl-2-oxabicyclo 3.3.0 octan-3-one, 3ar,4s,5r,6as-hexahydro-5-hydroxy-4-hydroxymethyl-2h-cyclopenta b furan-2-one, 3ar,4s,5r,6as-5-hydroxy-4-hydroxymethyl-3,3a,4,5,6,6a-hexahydrocyclopenta b furan-2-one, l-corey lactone
PubChem CID 2724453
IUPAC Name (3aR,4S,5R,6aS)-5-hydroxy-4-(hydroxymethyl)-3,3a,4,5,6,6a-hexahydrocyclopenta[b]furan-2-one
SMILES OC[C@H]1[C@H](O)C[C@@H]2OC(=O)C[C@H]12

Specifications

Melting Point 117°C to 119°C
Quantity 250 mg
Optical Rotation −44° (c=1.4 in Methanol)
Formula Weight 172.18
Percent Purity 98%
Chemical Name or Material (3aR,4S,5R,6aS)-(-)-Hexahydro-5-hydroxy-4-hydroxymethyl-2H-cyclopenta[b]furan-2-one
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only