missing translation for 'onlineSavingsMsg'
Learn More

3a,4,7,7a-Tetrahydroindene (stabilized with BHT) 98.0+%, TCI America™

Catalog No. T131525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T131525ML 25 mL
T1315500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T131525ML Supplier TCI America Supplier No. T131525ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3048-65-5
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
MDL Number MFCD00021227
InChI Key UFERIGCCDYCZLN-UHFFFAOYSA-N
Synonym 3a,4,7,7a-tetrahydroindene, 1h-indene, 3a,4,7,7a-tetrahydro, indene, 3a,4,7,7a-tetrahydro, 4,7,8,9-tetrahydroindene, bicyclo 4,3,0 nona-3,7-diene, tetrahydroindene russian, bicyclo 4.3.0 nona-3,7-diene, 1h-indene, 3a,4,7,7a-tetrahydro-, cis, cis-bicyclo 4.3.0 nona-3,7-diene, 1h-indene, 3a,4,7,7a-tetrahydro-, trans
PubChem CID 62475
IUPAC Name 3a,4,7,7a-tetrahydro-1H-indene
SMILES C1C=CCC2C1CC=C2

Specifications

Color Yellow
Boiling Point 160°C
Quantity 25 mL
UN Number 3295
Formula Weight 120.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3a,4,7,7a-Tetrahydroindene (stabilized with BHT)
EINECSNumber (4)-0581
RTECSNumber NK8750000
TSCA Yes