accessibility menu, dialog, popup

UserName

CAS RN 3048-65-5

CAS RN 3048-65-5

IUPAC Name: 3a,4,7,7a-tetrahydro-1H-indene
Synonyms: 3a,4,7,7a-tetrahydro-1H-indene
Molecular Weight (g/mol): 120.2
Molecular Formula: C9H12
InChi Key: UFERIGCCDYCZLN-UHFFFAOYNA-N
SMILES: C1C=CC2CC=CCC12
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results