Learn More
Greiner Bio-One 384-well Small Volume™ HiBase Polystyrene Microplates
GREENER_CHOICE
Features round wells with a conical geometry. Greiner Bio-One 384-Well Small Volume™ HiBase Polystyrene Microplates is free of detectable DNase, RNase, human DNA.
$1,476.00 - $5,854.11
Specifications
Format Standard
Array 16 x 24
Material Polystyrene
No. of Wells 384
View More Specs
Products 4
Catalog Number Mfr. No. Color Sterility No. per Case Price Quantity & Availability  
Catalog Number Mfr. No. Color Sterility No. per Case Price Quantity & Availability  
07000040
GREENER_CHOICE
View Documents Promotion Details
Greiner Bio-One
78407625
Black Non-sterile 150
Case of 150 Each for $5,747.04
Only null left
07000041
GREENER_CHOICE
View Documents Promotion Details
Greiner Bio-One
78407525
White Non-sterile 150
Case of 150 Each for $5,854.11
Only null left
07000065
GREENER_CHOICE
View Documents Promotion Details
Greiner Bio-One
784076
Black Non-sterile 40
Case of 40 Each for $1,541.64
Only null left
07000066
GREENER_CHOICE
View Documents Promotion Details
Greiner Bio-One
784075
White Non-sterile 40
Case of 40 Each for $1,476.00
Only null left
Description
Description
  • Non-Pyrogenic
  • Material: Polystyrene
  • Free of detectable DNase, RNase, human DNA
  • Traced way back to production through defined LOT number system
  • Working volume: 4μL - 25μL
Certifications
DIN ISO 9001
Specifications
Specifications
Standard
16 x 24
Polystyrene
384