missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 3500xL Dx Genetic Analyzer
GREENER_CHOICE

Catalog No. A27856
Encompass
Change view
Click to view available options
Quantity:
1 System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27856 1 System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A27856 Supplier Applied Biosystems™ Supplier No. A27856

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Content And Storage This system package consists of a 24-capillary instrument, Data Collection Software, and a Dell Workstation and monitor.
For Use With (Equipment) 3500xL Dx Genetic Analyzer
For Use With (Application) Sequencing
Product Type Genetic Analyzer
Quantity 1 System
Shipping Condition Room Temperature
Species Human