missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 3500xL Dx Data Collection Software 3, Upgrade Kit

Catalog No. A27859
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27859 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A27859 Supplier Applied Biosystems™ Supplier No. A27859

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

3500xL Dx Series Data Collection Software 3 provides the ideal features and functionality for use in both clinical and research environments.

3500xL Dx Series Data Collection Software 3 provides the ideal features and functionality for use in both clinical and research environments. Powered by the Windows™ 7 operating system, the 3500xL Dx Series Data Collection Software 3 provides:

  • Dual-mode user interface, the Diagnostic Mode and the Research Use Only Mode
  • Enables both fragment analysis and Sanger sequencing in the secure Diagnostic Mode
  • Offers superior flexibility for clinical research with the Research Use Only Mode
  • Enables export of validated assay protocols from Research Use Only Mode to Diagnostic Mode
  • Comprehensive dashboard that delivers regular updates of instrument status and remaining days of onboard consumables
  • SAE (Security, Audit, and Electronic) signature capability to assist with compliance with governing regulations

This product is a software upgrade package. It includes a computer system, monitor, 3500xL Dx Series Data Collection Software 3 upgrade USB flash drive, one licensing kit with single license code, and two user document USB flash drives.

For an upgrade, a full install and operational and performance qualification package by a certified engineer is required. These services are ordered and provided separately.

Specifications

Content And Storage • Dell Workstation and monitor
• 3500 Dx Series Data Collection Software 3 USB
• 1 licensing kit with single license code
• 2 user document USBs
Format Kit
For Use With (Equipment) 3500xL Dx Genetic Analyzer
For Use With (Application) Sequencing
Product Type Data Collection Software
Quantity 1 Kit
Shipping Condition Room Temperature