Learn More
Applied Biosystems™ 3500 Dx Genetic Analyzer
Specifications
Specifications
| Content And Storage | This system package consists of an 8-capillary instrument, Data Collection Software, and a Dell Workstation and monitor. |
| For Use With (Equipment) | 3500 Dx Genetic Analyzer |
| For Use With (Application) | Sequencing |
| Product Type | Genetic Analyzer |
| Quantity | 1 System |
| Shipping Condition | Room Temperature |
| Species | Human |
Frequently Asked Questions (FAQs)
Some of the other causes of an Unstable Electrophoresis current detected error message are:
1.Leak on the system
2. Polymer that:
-Has expired
-Has been left on the instrument for more than the recommended time
-Is a mixture of expired polymer and non-expired polymer
3. Running Buffer that was:
-On the instrument longer than 2 weeks
-Not filled to the fill line or evaporated below the fill line
4. An arcing event that was not cleaned afterwards using the water wash wizard
5. Not performing regular maintenance on the instrument
6. Hardware issues
Inspect the system for leaks. If you do not see any leaks on the system, perform the wash the pump chamber and channels wizard using the conditioning pouch and place fresh, non-expired polymer and fresh Anode and Cathode buffer containers. If the problem persists, a service call may be required.
If the instrument is not in use, it is not necessary to replace the buffer every 14 days. However, the buffer level needs to remain at the fill line to prevent the capillary or array from drying out. Top off the buffer volume with water if necessary. Replace the anode and cathode buffer containers prior to starting a new run.