missing translation for 'onlineSavingsMsg'
Learn More

CDS Empore™ 3290 Strontium RAD DISK, 47 mm

Selectively bonding Sr2+ cations from sample solutions. Used for nucleid species detection. Reduce extraction time and solvent use for analytical preparation of aqueous environmental samples.

Supplier:  CDS Empore™ 98040500641EA

 View more versions of this product

Catalog No. 13-110-075

Only null left
Explore available promotions

Description

Description

Empore™ RAD Disks provide an efficient alternative to conventional radiochemical sample preparation methods that use wet chemistry or packed columns.

  • Empore RAD disks provide a sample prep solution for large volume aqueous samples
  • The disk format provides a large surface area for sorbent/sample contact
  • Fast flow rates and high throughput may be realized with use of an Empore RAD disk
  • RAD disks form convenient direct counting sources when used in applicable test methods
  • Available with a variety of packings in 47 mm size
  • Approved to detect Strontium-90 in DOE Method RP515
  • Total Sr amount: not exceed 3 mg
  • Method detection limit: 1 pCi/L for Strontium-90
Specifications

Specifications

530 mg
Vacuum Manifolds or Automated SPE Equipment
Extraction Disk
Disk
12 μm
20/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

To reduce the risks associated with environmental contamination, handling radiochemical materials or false negative results: Read, understand and follow all recommendations in the product MSDS and Instructions for Use.

To reduce the risks associated with radiochemical materials: Always wear the proper PPE (Personal Protection Equipment) when handling used product.

To reduce the risks associated with false negative results for radiochemicals: Always use the Empore™ Cesium Rad Disks by the expiration date.

To reduce the risks associated with environmental contamination: Dispose of used disks according to standard radiochemical procedures and regulations. Unused disks may be disposed of in accordance with standard procedure for laboratory waste.