missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 310 Computer Kit (Windows™) OS

Catalog No. 4327282
Encompass
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4327282 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4327282 Supplier Applied Biosystems™ Supplier No. 4327282

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This kit includes a Dell™ computer and monitor with the Windows™ operating system, intended for the 310 Genetic Analyzer.

This kit includes a Dell™ computer and monitor with the Windows™ operating system, intended for the 310 Genetic Analyzer.

This kit does not include any Data Collection or Analysis software.

Specifications

Content And Storage Includes a Dell™ computer and monitor with the Windows™ operating system.
Format Kit
For Use With (Equipment) 310 Genetic Analyzer
For Use With (Application) Sequencing
Product Type Computer Kit
Quantity Each

For Research Use Only. Not for use in diagnostic procedures.