missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 300 μL PolySpring Autosampler Vial Inserts

Catalog No. 13622217
Change view
Click to view available options
Material:
Glass, Polypropylene
Polypropylene
Volume (Metric) Usable:
250 μL
0.3 mL
Volume (Metric) Total:
325 μL
0.375 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Volume (Metric) Usable Volume (Metric) Total
13622217 Polypropylene 250 μL 325 μL
0339135 Glass, Polypropylene 0.3 mL 0.375 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13622217 Supplier Fisherbrand™ Supplier No. 13622217
Get 1 Extra Pack of Vials When You Buy 2
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fisherbrand™ PolySpring inserts are available in glass or polypropylene and are compatible with 9 mm HPLC and GC autosampler vials.

Nominal O.D. of 6 mm to fit Target wide-opening vials.

Specifications

Color Clear
Diameter (Metric) Outer 6 mm
Material Polypropylene
Volume (Metric) 300 μL
No. per Pack 100
Volume (Metric) Residual < 2 μL
Base Style Conical
Product Line Fisherbrand
Volume (Metric) Total 325 μL
Volume (Metric) Usable 250 μL
Height (Metric) 30 mm
Product Type Autosampler Vial Spring Insert
Show More Show Less