missing translation for 'onlineSavingsMsg'
Learn More

Troemner™ 300 g Electronic Balance UltraClass Weight

Catalog No. 02218086
Change view
Click to view available options
Certifications/Compliance:
Traceable Certificate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
02218086 Traceable Certificate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02218086 Supplier Troemner™ Supplier No. 30390314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Combine high precision with the advantage of two-piece construction. UltraClass weights, exclusive to Troemner™ are the most precise two-piece weights available with weight tolerances that equal ANSI/ASTM E617 Class 0 or OIML R 111 Class E2 tolerances.

  • Used in extremely stable environments
  • Supplied in a rugged, precision-fit, high quality polycarbonate case with handling glove

Specifications

Product Type Two-Piece Analytical Weight
Certifications/Compliance Traceable Certificate
Classification UltraClass
Material Stainless Steel
Case Material Polycarbonate
Density 7.95 g/cm3 at 20°C
Mass 300 g
Tolerance 0.38 mg
Length (Metric) 8.89 cm
Height (Metric) Case 11.43 cm
For Use With (Application) Reference standard calibration
For Use With (Equipment) Calibration of high-precision analytical balances with readability as low as 0.01 mg
Includes Cylindrical Weight, Polycarbonate Case, Glove
Quantity Each
Show More Show Less