missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Vinylpyridine (stabilized with TBC) 96.0+%, TCI America™
$365.35 - $1094.21
Chemical Identifiers
| CAS | 1121-55-7 |
|---|---|
| Molecular Formula | C7H7N |
| Molecular Weight (g/mol) | 105.14 |
| MDL Number | MFCD02178019 |
| InChI Key | DPZYLEIWHTWHCU-UHFFFAOYSA-N |
| PubChem CID | 14272 |
| IUPAC Name | 3-ethenylpyridine |
| SMILES | C=CC1=CC=CN=C1 |
Chemical Identifiers
| 1121-55-7 | |
| 105.14 | |
| DPZYLEIWHTWHCU-UHFFFAOYSA-N | |
| 3-ethenylpyridine |
| C7H7N | |
| MFCD02178019 | |
| 14272 | |
| C=CC1=CC=CN=C1 |
Specifications
| 1121-55-7 | |
| 82°C | |
| MFCD02178019 | |
| 3073 | |
| C=CC1=CC=CN=C1 | |
| 105.14 | |
| 105.14 | |
| Liquid |
| Yellow | |
| C7H7N | |
| 1 g | |
| DPZYLEIWHTWHCU-UHFFFAOYSA-N | |
| 3-ethenylpyridine | |
| 14272 | |
| ≥96.0% (GC,T) | |
| 3-Vinylpyridine (stabilized with TBC) |
Safety and Handling
TSCA : Yes
Recommended Storage : Freezer