missing translation for 'onlineSavingsMsg'
Learn More

3-Vinylpyridine (stabilized with TBC) 96.0+%, TCI America™

Catalog No. V00995G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V00995G 5 g
V00991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V00995G Supplier TCI America Supplier No. V00995G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1121-55-7
Molecular Formula C7H7N
Molecular Weight (g/mol) 105.14
MDL Number MFCD02178019
InChI Key DPZYLEIWHTWHCU-UHFFFAOYSA-N
PubChem CID 14272
IUPAC Name 3-ethenylpyridine
SMILES C=CC1=CC=CN=C1

Specifications

Color Yellow
Boiling Point 82°C
Quantity 5 g
UN Number 3073
Formula Weight 105.14
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Vinylpyridine (stabilized with TBC)
TSCA Yes
Recommended Storage Freezer