missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Vinylpyridine (stabilized with TBC) 96.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 82°C |
| Quantity | 1 g |
| UN Number | 3073 |
| Formula Weight | 105.14 |
| Percent Purity | ≥96.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Vinylpyridine (stabilized with TBC) |
Safety and Handling
| TSCA | Yes |
| Recommended Storage | Freezer |