missing translation for 'onlineSavingsMsg'
Learn More

3-Vinylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. V01411G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V01411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V01411G Supplier TCI America Supplier No. V01411G

Specifications

Chemical Name or Material 3-Vinylphenylboronic Acid (contains varying amounts of Anhydride)
CAS 15016-43-0
Melting Point 145°C
Color White
Molecular Formula C8H9BO2
Quantity 1 g
MDL Number MFCD01074679
Synonym 3-vinylphenylboronic acid, 3-vinylphenyl boronic acid, 3-vinylbenzeneboronic acid, styrene-3-boronic acid, 3-ethenylphenyl boronic acid, m-vinylbenzeneboronic acid, boronic acid, 3-ethenylphenyl, 3-vinylphenylbronicacid, 3-vinyl-phenylboronic acid
InChI Key SYBQEKBVWDPVJM-UHFFFAOYSA-N
SMILES B(C1=CC(=CC=C1)C=C)(O)O
IUPAC Name (3-ethenylphenyl)boronic acid
Molecular Weight (g/mol) 147.968
PubChem CID 4366886
Formula Weight 147.97
Physical Form Crystalline Powder
Show More Show Less