missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Undecylthiophene 98.0+%, TCI America™
$270.29 - $784.76
Chemical Identifiers
| CAS | 129607-86-9 |
|---|---|
| Molecular Formula | C15H26S |
| Molecular Weight (g/mol) | 238.433 |
| MDL Number | MFCD00130144 |
| InChI Key | STIIRMZYURVVGK-UHFFFAOYSA-N |
| PubChem CID | 566841 |
| IUPAC Name | 3-undecylthiophene |
| SMILES | CCCCCCCCCCCC1=CSC=C1 |
Chemical Identifiers
| 129607-86-9 | |
| 238.433 | |
| STIIRMZYURVVGK-UHFFFAOYSA-N | |
| 3-undecylthiophene |
| C15H26S | |
| MFCD00130144 | |
| 566841 | |
| CCCCCCCCCCCC1=CSC=C1 |
Specifications
| 129607-86-9 | |
| 166°C | |
| MFCD00130144 | |
| STIIRMZYURVVGK-UHFFFAOYSA-N | |
| 3-undecylthiophene | |
| 566841 | |
| ≥98.0% (GC) | |
| 3-Undecylthiophene |
| Yellow | |
| C15H26S | |
| 1 g | |
| CCCCCCCCCCCC1=CSC=C1 | |
| 238.433 | |
| 238.43 | |
| Liquid |
Safety and Handling
TSCA : No