accessibility menu, dialog, popup

UserName

H3CS

CAS RN 129607-86-9

IUPAC Name: 3-undecylthiophene
Synonyms: 3-undecylthiophene
Molecular Weight (g/mol): 238.43
Molecular Formula: C15H26S
InChi Key: STIIRMZYURVVGK-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC1=CSC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results