missing translation for 'onlineSavingsMsg'
Learn More

3-Tropanylindole-3-carboxylate hydrochloride, Thermo Scientific Chemicals

Catalog No. AAJ62807MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62807MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62807MC Supplier Thermo Scientific Chemicals Supplier No. J62807MC

A selective 5-HT3 serotonin receptor antagonist

A selective 5-HT3 serotonin receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105826-92-4
Molecular Formula C17H21ClN2O2
Molecular Weight (g/mol) 320.817
MDL Number MFCD00210221
InChI Key XIEGSJAEZIGKSA-ICHLHHFLSA-N
Synonym sdz-ics 930
PubChem CID 71300439
IUPAC Name [(5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 1H-indole-3-carboxylate;hydrochloride
SMILES CN1C2CCC1CC(C2)OC(=O)C3=CNC4=CC=CC=C43.Cl

Specifications

Color White
Quantity 100 mg
UN Number UN2811
Merck Index 14,9783
Solubility Information Soluble in water
Formula Weight 320.82
Physical Form Powder
Chemical Name or Material 3-Tropanylindole-3-carboxylate hydrochloride
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber NL6012520
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only