missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Trimethylsilylpropynal 97.0+%, TCI America™
$333.67 - $993.54
Chemical Identifiers
| CAS | 2975-46-4 |
|---|---|
| Molecular Formula | C6H10OSi |
| Molecular Weight (g/mol) | 126.23 |
| MDL Number | MFCD01103709 |
| InChI Key | LJRWLSKYGWLYIM-UHFFFAOYSA-N |
| Synonym | 3-trimethylsilylpropynal, 3-trimethylsilyl-2-propynal, 3-trimethylsilyl propiolaldehyde, 3-trimethylsilyl propynal, trimethylsilyl propiolaldehyde, 2-propynal, 3-trimethylsilyl, trimethylsilylpropynal, 3-trimethylsilyl prop-2-ynal |
| PubChem CID | 2760865 |
| IUPAC Name | 3-(trimethylsilyl)prop-2-ynal |
| SMILES | C[Si](C)(C)C#CC=O |
Chemical Identifiers
| 2975-46-4 | |
| 126.23 | |
| LJRWLSKYGWLYIM-UHFFFAOYSA-N | |
| 2760865 | |
| C[Si](C)(C)C#CC=O |
| C6H10OSi | |
| MFCD01103709 | |
| 3-trimethylsilylpropynal, 3-trimethylsilyl-2-propynal, 3-trimethylsilyl propiolaldehyde, 3-trimethylsilyl propynal, trimethylsilyl propiolaldehyde, 2-propynal, 3-trimethylsilyl, trimethylsilylpropynal, 3-trimethylsilyl prop-2-ynal | |
| 3-(trimethylsilyl)prop-2-ynal |
Specifications
| 2975-46-4 | |
| 45°C | |
| MFCD01103709 | |
| 1993 | |
| LJRWLSKYGWLYIM-UHFFFAOYSA-N | |
| 3-(trimethylsilyl)prop-2-ynal | |
| 2760865 | |
| ≥97.0% (GC) | |
| 3-Trimethylsilylpropynal |
| Yellow | |
| C6H10OSi | |
| 1 g | |
| 3-trimethylsilylpropynal, 3-trimethylsilyl-2-propynal, 3-trimethylsilyl propiolaldehyde, 3-trimethylsilyl propynal, trimethylsilyl propiolaldehyde, 2-propynal, 3-trimethylsilyl, trimethylsilylpropynal, 3-trimethylsilyl prop-2-ynal | |
| C[Si](C)(C)C#CC=O | |
| 126.23 | |
| 126.23 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator