accessibility menu, dialog, popup

UserName

CAS RN 2975-46-4

CH3SiCH3CH3O

CAS RN 2975-46-4

IUPAC Name: 3-(trimethylsilyl)prop-2-ynal
Synonyms: 3-(trimethylsilyl)prop-2-ynal
Molecular Weight (g/mol): 126.23
Molecular Formula: C6H10OSi
InChi Key: LJRWLSKYGWLYIM-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CC=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)