missing translation for 'onlineSavingsMsg'
Learn More

3-Trimethylsilylpropynal 97.0+%, TCI America™

Catalog No. T29075G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29075G 5 g
T29071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29075G Supplier TCI America Supplier No. T29075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2975-46-4
Molecular Formula C6H10OSi
Molecular Weight (g/mol) 126.23
MDL Number MFCD01103709
InChI Key LJRWLSKYGWLYIM-UHFFFAOYSA-N
Synonym 3-trimethylsilylpropynal, 3-trimethylsilyl-2-propynal, 3-trimethylsilyl propiolaldehyde, 3-trimethylsilyl propynal, trimethylsilyl propiolaldehyde, 2-propynal, 3-trimethylsilyl, trimethylsilylpropynal, 3-trimethylsilyl prop-2-ynal
PubChem CID 2760865
IUPAC Name 3-(trimethylsilyl)prop-2-ynal
SMILES C[Si](C)(C)C#CC=O

Specifications

Color Yellow
Boiling Point 45°C
Quantity 5 g
UN Number 1993
Formula Weight 126.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Trimethylsilylpropynal
TSCA No
Recommended Storage Refrigerator